missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylcyanamide 98.0+%, TCI America™

Catalog No. D49525ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49525ML 5 mL
D49521ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49525ML Supplier TCI America Supplier No. D49525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3085-76-5
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008865
InChI Key DGCUISYKMONQDH-UHFFFAOYSA-N
PubChem CID 76520
IUPAC Name cyanobis(propan-2-yl)amine
SMILES CC(C)N(C#N)C(C)C

Specifications

Melting Point -27°C
Color Yellow
Boiling Point 94°C
Quantity 5 mL
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropylcyanamide