accessibility menu, dialog, popup

UserName

H3CCH3NNCH3H3C

CAS RN 3085-76-5

IUPAC Name: cyanobis(propan-2-yl)amine
Synonyms: cyanodiisopropylamine
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: DGCUISYKMONQDH-UHFFFAOYSA-N
SMILES: CC(C)N(C#N)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results