missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine Hydrochloride 99.0+%, TCI America™

Catalog No. D092625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D092625G 25 g
D0926500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D092625G Supplier TCI America Supplier No. D092625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 819-79-4
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.651
MDL Number MFCD00034860
InChI Key URAZVWXGWMBUGJ-UHFFFAOYSA-N
PubChem CID 11320964
IUPAC Name N-propan-2-ylpropan-2-amine;hydrochloride
SMILES CC(C)NC(C)C.Cl

Specifications

Melting Point 217°C
Color White
Quantity 25 g
Formula Weight 137.65
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diisopropylamine Hydrochloride
EINECSNumber (2)-0136
RTECSNumber IM4340000
TSCA No