missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropylamine, 99%, Spectrum™ Chemical
$1,081.68 - $1,081.68
Chemical Identifiers
| CAS | 108-18-9 |
|---|---|
| Molecular Formula | C6H15N |
| Molecular Weight (g/mol) | 101.19 |
| InChI Key | UAOMVDZJSHZZME-UHFFFAOYSA-N |
| IUPAC Name | bis(propan-2-yl)amine |
| SMILES | CC(C)NC(C)C |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.