missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl sulfide, 99%

Catalog No. AAB2130209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2130209 10 g
AAB2130218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2130209 Supplier Thermo Scientific Chemicals Supplier No. B2130209

CAS: 625-80-9 | C6H14S | 118.24 g/mol

Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 625-80-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00008895
InChI Key XYWDPYKBIRQXQS-UHFFFAOYSA-N
Synonym diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
PubChem CID 12264
SMILES CC(C)SC(C)C

Specifications

Density 0.814
Boiling Point 119°C to 120°C
Flash Point 7°C (44°F)
Odor Stench
Refractive Index 1.4385
Linear Formula (CH3)2CHSCH(CH3)2
Quantity 10 g
UN Number UN1993
Beilstein 1731257
Solubility Information Not miscible or difficult to mix with water.
IUPAC Name 2-propan-2-ylsulfanylpropane
Formula Weight 118.24
Percent Purity 99%
Chemical Name or Material Diisopropyl sulfide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-911-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only