missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Sulfate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | -19°C |
| Color | Yellow |
| Boiling Point | 94°C |
| Quantity | 5 g |
| UN Number | 1760 |
| Formula Weight | 182.23 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diisopropyl Sulfate |
Safety and Handling
| EINECSNumber | (2)-1673 |
| RTECSNumber | WS8050000 |
| TSCA | No |
| Recommended Storage | Freezer |