accessibility menu, dialog, popup

UserName

H3CCH3OSOOOCH3H3C

CAS RN 2973-10-6

IUPAC Name: bis(propan-2-yl) sulfate
Synonyms: diisopropyl sulfate
Molecular Weight (g/mol): 182.23
Molecular Formula: C6H14O4S
InChi Key: HWBLTYHIEYOAOL-UHFFFAOYSA-N
SMILES: CC(C)OS(=O)(=O)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results