missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl methylphosphonate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1290422

 View more versions of this product

Catalog No. AAA1290422

Only null left
Explore available promotions

Description

Description

Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

Solubility
Miscible with water.

Notes
Store in cool place. Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, reducing agents, strong reducing agents and phosphorus halides.
Specifications

Specifications

Diisopropyl methylphosphonate
0.976
C7H17O3P
MFCD00015022
1761635
Miscible with water.
CC(C)OP(C)(=O)OC(C)C
180.18
CHEBI:77325
95%
1445-75-6
50°C to 51°C (1 mmHg)
1.41
100 g
diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
WOAFDHWYKSOANX-UHFFFAOYSA-N
2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
3073
180.18

Chemical Identifiers

1445-75-6
180.18
WOAFDHWYKSOANX-UHFFFAOYSA-N
3073
2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
C7H17O3P
MFCD00015022
diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
CHEBI:77325
CC(C)OP(C)(=O)OC(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 215-896-0

RTECSNumber : SZ9090000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only