missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl methylphosphonate, 95%, Thermo Scientific Chemicals

Catalog No. AAA1290406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290406 5 g
AAA1290414 25 g
AAA1290422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1290406 Supplier Thermo Scientific Chemicals Supplier No. A1290406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl methylphosphonate is used as a precursor to prepare beta-keto phosphonates. It acts as an intermediate for Horner-Wadsworth-Emmons olefination of carbonyl compounds.

Solubility
Miscible with water.

Notes
Store in cool place. Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, reducing agents, strong reducing agents and phosphorus halides.

Chemical Identifiers

CAS 1445-75-6
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.18
MDL Number MFCD00015022
InChI Key WOAFDHWYKSOANX-UHFFFAOYSA-N
Synonym diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
PubChem CID 3073
ChEBI CHEBI:77325
IUPAC Name 2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(C)(=O)OC(C)C

Specifications

Density 0.976
Boiling Point 50°C to 51°C (1 mmHg)
Refractive Index 1.41
Quantity 5 g
Beilstein 1761635
Solubility Information Miscible with water.
Formula Weight 180.18
Percent Purity 95%
Chemical Name or Material Diisopropyl methylphosphonate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-896-0
RTECSNumber SZ9090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only