missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Malonate 99.0+%, TCI America™
$54.06 - $212.51
Chemical Identifiers
| CAS | 13195-64-7 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.223 |
| MDL Number | MFCD00059359 |
| InChI Key | QRVSDVDFJFKYKA-UHFFFAOYSA-N |
| Synonym | Malonic Acid Diisopropyl Ester |
| PubChem CID | 83223 |
| IUPAC Name | dipropan-2-yl propanedioate |
| SMILES | CC(C)OC(=O)CC(=O)OC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M064825ML
|
TCI America
M064825ML |
25 mL |
Each for $54.06
|
|
|||||
|
M0648500ML
|
TCI America
M0648500ML |
500 mL |
Each for $212.51
|
|
|||||
Chemical Identifiers
| 13195-64-7 | |
| 188.223 | |
| QRVSDVDFJFKYKA-UHFFFAOYSA-N | |
| 83223 | |
| CC(C)OC(=O)CC(=O)OC(C)C |
| C9H16O4 | |
| MFCD00059359 | |
| Malonic Acid Diisopropyl Ester | |
| dipropan-2-yl propanedioate |
Specifications
| 13195-64-7 | |
| Colorless | |
| C9H16O4 | |
| 25 mL | |
| QRVSDVDFJFKYKA-UHFFFAOYSA-N | |
| dipropan-2-yl propanedioate | |
| 83223 | |
| ≥99.0% (GC) | |
| Diisopropyl Malonate |
| -51°C | |
| 106°C | |
| MFCD00059359 | |
| Malonic Acid Diisopropyl Ester | |
| CC(C)OC(=O)CC(=O)OC(C)C | |
| 188.223 | |
| 188.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2549
TSCA : No