missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Malonate 99.0+%, TCI America™
Supplier: TCI America M064825ML
Specifications
| Diisopropyl Malonate | |
| -51°C | |
| 106°C | |
| MFCD00059359 | |
| Malonic Acid Diisopropyl Ester | |
| CC(C)OC(=O)CC(=O)OC(C)C | |
| 188.223 | |
| 188.22 | |
| Liquid |
| 13195-64-7 | |
| Colorless | |
| C9H16O4 | |
| 25 mL | |
| QRVSDVDFJFKYKA-UHFFFAOYSA-N | |
| dipropan-2-yl propanedioate | |
| 83223 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 13195-64-7 | |
| 188.223 | |
| QRVSDVDFJFKYKA-UHFFFAOYSA-N | |
| 83223 | |
| CC(C)OC(=O)CC(=O)OC(C)C |
| C9H16O4 | |
| MFCD00059359 | |
| Malonic Acid Diisopropyl Ester | |
| dipropan-2-yl propanedioate |
Safety and Handling
EINECSNumber : (2)-2549
TSCA : No