missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ketone, 98%

Catalog No. AAA1287536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1287536 500 g
AAA1287522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1287536 Supplier Thermo Scientific Chemicals Supplier No. A1287536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 565-80-0 | C7H14O | 114.188 g/mol

2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

Melting Point -69°C
Density 0.803
Boiling Point 124°C to 125°C
Flash Point 15°C (59°F)
Refractive Index 1.399
Quantity 500 g
UN Number UN1224
Beilstein 773782
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material Diisopropyl ketone
Show More Show Less
Hazard Category H225-H332
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 209-294-7
RTECSNumber SA8575500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only