accessibility menu, dialog, popup

UserName

CAS RN 565-80-0

IUPAC Name: 2,4-dimethylpentan-3-one
Synonyms: 2,4-dimethyl-3-pentanone
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: HXVNBWAKAOHACI-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)