missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl bromomethylphosphonate, 97%

Catalog No. p-7046270
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1376306 5 g
AAL1376314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1376306 Supplier Thermo Scientific Chemicals Supplier No. L1376306

CAS: 98432-80-5 | C7H16BrO3P | 259.08 g/mol

Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 98432-80-5
Molecular Formula C7H16BrO3P
Molecular Weight (g/mol) 259.08
MDL Number MFCD00236036
InChI Key JNONGIUWXHOEEN-UHFFFAOYSA-N
Synonym diisopropyl bromomethylphosphonate, bromomethylphosphonic acid diisopropyl ester, diisopropyl bromomethyl phosphonate, bromomethylphosphonate diisopropyl ester, 2-bromomethyl propan-2-yloxy phosphoryl oxypropane
PubChem CID 4366885
IUPAC Name 2-[bromomethyl(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(=O)(CBr)OC(C)C

Specifications

Boiling Point 80°C (1 mmHg)
Refractive Index 1.4495
Quantity 5 g
UN Number UN3265
Beilstein 4740388
Solubility Information Insoluble in water.
Formula Weight 259.09
Percent Purity 97%
Chemical Name or Material Diisopropyl bromomethylphosphonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only