CAS RN 98432-80-5
IUPAC Name:
bis(propan-2-yl) (bromomethyl)phosphonate
Synonyms:
diisopropyl bromomethylphosphonate
Molecular Weight (g/mol):
259.08
Molecular Formula:
C7H16BrO3P
InChi Key:
JNONGIUWXHOEEN-UHFFFAOYSA-N
SMILES:
CC(C)OP(=O)(CBr)OC(C)C
1
–
1
of
1
results
1
Promotions Available
Diisopropyl bromomethylphosphonate, 97%
Diisopropyl bromomethylphosphonate, 97%
CAS: 98432-80-5 Molecular Formula: C7H16BrO3P Molecular Weight (g/mol): 259.08 MDL Number: MFCD00236036 InChI Key: JNONGIUWXHOEEN-UHFFFAOYSA-N Synonym: diisopropyl bromomethylphosphonate,bromomethylphosphonic acid diisopropyl ester,diisopropyl bromomethyl phosphonate,bromomethylphosphonate diisopropyl ester,2-bromomethyl propan-2-yloxy phosphoryl oxypropane PubChem CID: 4366885 IUPAC Name: 2-[bromomethyl(propan-2-yloxy)phosphoryl]oxypropane SMILES: CC(C)OP(=O)(CBr)OC(C)C