missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl (Bromomethyl)boronate 95.0+%, TCI America™

Catalog No. D36495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36495G 5 g
D36491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36495G Supplier TCI America Supplier No. D36495G
Only null left

Chemical Identifiers

CAS 137297-49-5
Molecular Formula C7H16BBrO2
Molecular Weight (g/mol) 222.917
MDL Number MFCD01631222
InChI Key FNHYYZNZIWONBQ-UHFFFAOYSA-N
Synonym (Bromomethyl)boronic Acid Diisopropyl Ester
PubChem CID 2762526
IUPAC Name bromomethyl-di(propan-2-yloxy)borane
SMILES B(CBr)(OC(C)C)OC(C)C

Specifications

Color Yellow
Quantity 5 g
UN Number 3272
Formula Weight 222.92
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl (Bromomethyl)boronate
TSCA No
Recommended Storage Refrigerator