accessibility menu, dialog, popup

UserName

BBrOH3CCH3OCH3CH3

CAS RN 137297-49-5

IUPAC Name: bis(propan-2-yl) (bromomethyl)boronate
Synonyms: diisopropyl bromomethylboronate
Molecular Weight (g/mol): 222.92
Molecular Formula: C7H16BBrO2
InChi Key: FNHYYZNZIWONBQ-UHFFFAOYSA-N
SMILES: CC(C)OB(CBr)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results