Learn More
Diisopropyl azodicarboxylate, 94%
CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol
Supplier: Thermo Scientific Chemicals 327560250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiatorsSpecifications
| Diisopropyl azodicarboxylate | |
| 2446-83-5 | |
| Orange-Yellow to Red | |
| 75.0°C (0.2 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4180 to 1.4220 | |
| MFCD00008875 | |
| 17,39 | |
| 15,3217 | |
| Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
| CC(C)OC(=O)N=NC(=O)OC(C)C | |
| 202.21 | |
| 202.21 | |
| Liquid |
| 94% | |
| 3.0°C to 5.0°C | |
| 1.0200g/mL | |
| 106°C | |
| 92% min. (GC) | |
| C8H14N2O4 | |
| (CH3)2CHO2CN=NCO2CH(CH3)2 | |
| 25 g | |
| 1.02 | |
| z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide | |
| VVWRJUBEIPHGQF-KTKRTIGZSA-N | |
| propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate | |
| 56776520 | |
| 94% |
Chemical Identifiers
| 2446-83-5 | |
| 202.21 | |
| VVWRJUBEIPHGQF-KTKRTIGZSA-N | |
| 56776520 | |
| CC(C)OC(=O)N=NC(=O)OC(C)C |
| C8H14N2O4 | |
| MFCD00008875 | |
| z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide | |
| propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.<br
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 219-502-8
RUO – Research Use Only