accessibility menu, dialog, popup

UserName

CH3H3COONNOOCH3H3C(E/Z)

CAS RN 2446-83-5

IUPAC Name: N-{[(propan-2-yloxy)carbonyl]imino}(propan-2-yloxy)formamide
Synonyms: diisopropyl azodicarboxylate
Molecular Weight (g/mol): 202.21
Molecular Formula: C8H14N2O4
InChi Key: VVWRJUBEIPHGQF-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)N=NC(=O)OC(C)C
Boiling Point 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)