missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl azodicarboxylate, 94%

Catalog No. AC327565000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327565000 500 g
AC327560050 5 g
AC327560250 25 g
AC327561000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC327565000 Supplier Thermo Scientific Chemicals Supplier No. 327565000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators

Chemical Identifiers

CAS 2446-83-5
Molecular Formula C8H14N2O4
Molecular Weight (g/mol) 202.21
MDL Number MFCD00008875
InChI Key VVWRJUBEIPHGQF-KTKRTIGZSA-N
Synonym z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide
PubChem CID 56776520
IUPAC Name propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate
SMILES CC(C)OC(=O)N=NC(=O)OC(C)C

Specifications

Melting Point 3.0°C to 5.0°C
Color Orange-Yellow to Red
Density 1.0200g/mL
Boiling Point 75.0°C (0.2 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Linear Formula (CH3)2CHO2CN=NCO2CH(CH3)2
Refractive Index 1.4180 to 1.4220
Quantity 500 g
Fieser 17,39
Specific Gravity 1.02
Merck Index 15,3217
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 202.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Diisopropyl azodicarboxylate, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.<br
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 219-502-8

RUO – Research Use Only