missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine, Spectrum™ Chemical

Catalog No. 18610202
Change view
Click to view available options
Quantity:
500 g
2.5 kg
40 lb.
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610202 2.5 kg Poly Pail
18610204 500 g Poly Pail
18610203 40 lb. Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610202 Supplier Spectrum Chemical Mfg Cor Supplier No. D13242.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 110-97-4

Specifications

CAS Max % 1
Boiling Point 249°C / 480.2°F
Packaging Poly Pail
Quantity 2.5 kg
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Diisopropanolamine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.