missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine, 98+%, sum of isomers

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol

Supplier:  Thermo Scientific Chemicals A1845130

 View more versions of this product

Catalog No. AAA1845130


Only null left
Explore available promotions

Description

Description

Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

Solubility
Soluble in water 20°C 870g/L. Miscible in alcohol.

Notes
Air Sensitive & Hygroscopic. Store away from oxidizing agents, air, water/moisture and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

Diisopropanolamine
110-97-4
1.004
130°C (266°F)
C6H15NO2
250 g
Air sensitive; Hygroscopic
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
LVTYICIALWPMFW-UHFFFAOYNA-N
1-[(2-hydroxypropyl)amino]propan-2-ol
8086
≥98%
sum of isomers
38°C to 45°C
249°C to 250°C
Amine-like
MFCD00004531
605363
14,3195
Soluble in water 20°C 870g/L. Miscible in alcohol.
CC(O)CNCC(C)O
133.19
133.19

Chemical Identifiers

110-97-4
133.19
LVTYICIALWPMFW-UHFFFAOYNA-N
8086
CC(O)CNCC(C)O
C6H15NO2
MFCD00004531
diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
1-[(2-hydroxypropyl)amino]propan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 203-820-9

RTECSNumber : UB6600000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only