missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopinocampheylchloroborane, 1.8M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog No. p-3586320
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388421000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC388421000 Supplier Thermo Scientific Chemicals Supplier No. 388421000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112246-73-8 | Molecular Formula: C20H34BCl | Molecular Weight: 320.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 112246-73-8 , 80-56-8 , 71-43-2 , 64742-89-8 , 64742-89-8
Molecular Formula C20H34BCl
Molecular Weight (g/mol) 320.75
MDL Number MFCD00074808
InChI Key PSEHHVRCDVOTID-NAVXHOJHSA-N
Synonym +-b-chlorodiisopinocamphenylborane
PubChem CID 133124869
IUPAC Name chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
SMILES C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

Specifications

Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 320.77
Chemical Name or Material (+)-Diisopinocampheylchloroborane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways. Suspected of damaging fertility. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only