accessibility menu, dialog, popup

UserName

CAS RN 112246-73-8

IUPAC Name: chlorobis[(1S,2R,3S,5S)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl]borane
Synonyms: chlorobis[(1S,2R,3S,5S)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl]borane
Molecular Weight (g/mol): 320.75
Molecular Formula: C20H34BCl
InChi Key: PSEHHVRCDVOTID-NAVXHOJHSA-N
SMILES: C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results