missing translation for 'onlineSavingsMsg'
Learn More

Diisopentyl ether, mixture of isomers, 96%

CAS: 544-01-4 | C10H22O | 158.285 g/mol

Supplier:  Thermo Scientific Chemicals A11231AE

 View more versions of this product

Catalog No. AAA11231AE


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diisopentyl ether
544-01-4
172°C to 174°C
C10H22O
MFCD00008947
UN3271
14,5118
AQZGPSLYZOOYQP-UHFFFAOYSA-N
3-methyl-1-(3-methylbutoxy)butane
10989
96%
Mixture of isomers
0.78
46°C (115°F)
1.407
100 mL
1698014
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
CC(C)CCOCCC(C)C
158.285
158.29

Chemical Identifiers

544-01-4
158.285
AQZGPSLYZOOYQP-UHFFFAOYSA-N
10989
CC(C)CCOCCC(C)C
C10H22O
MFCD00008947
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
3-methyl-1-(3-methylbutoxy)butane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 208-857-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only