missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisodecyl Adipate, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 240°C |
| Quantity | 500 mL |
| Formula Weight | 426.68 |
| Physical Form | Liquid |
| Chemical Name or Material | Diisodecyl Adipate |
Safety and Handling
| EINECSNumber | (2)-0861&(2)-0879 |
| RTECSNumber | AV1485000 |
| TSCA | Yes |