accessibility menu, dialog, popup

UserName

H3CCH3OOOOCH3CH3

CAS RN 27178-16-1

IUPAC Name: 1,6-bis(8-methylnonyl) hexanedioate
Synonyms: diisodecyl adipate
Molecular Weight (g/mol): 426.68
Molecular Formula: C26H50O4
InChi Key: YKGYQYOQRGPFTO-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCCOC(=O)CCCCC(=O)OCCCCCCCC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results