missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyldimethoxysilane 98.0+%, TCI America™
$37.29 - $95.06
Chemical Identifiers
| CAS | 17980-32-4 |
|---|---|
| Molecular Formula | C10H24O2Si |
| Molecular Weight (g/mol) | 204.385 |
| MDL Number | MFCD00236510 |
| InChI Key | NHYFIJRXGOQNFS-UHFFFAOYSA-N |
| PubChem CID | 3084284 |
| IUPAC Name | dimethoxy-bis(2-methylpropyl)silane |
| SMILES | CC(C)C[Si](CC(C)C)(OC)OC |
Chemical Identifiers
| 17980-32-4 | |
| 204.385 | |
| NHYFIJRXGOQNFS-UHFFFAOYSA-N | |
| dimethoxy-bis(2-methylpropyl)silane |
| C10H24O2Si | |
| MFCD00236510 | |
| 3084284 | |
| CC(C)C[Si](CC(C)C)(OC)OC |
Specifications
| 17980-32-4 | |
| 72°C | |
| MFCD00236510 | |
| NHYFIJRXGOQNFS-UHFFFAOYSA-N | |
| dimethoxy-bis(2-methylpropyl)silane | |
| 3084284 | |
| ≥98.0% (GC) | |
| Diisobutyldimethoxysilane |
| Colorless | |
| C10H24O2Si | |
| 5 mL | |
| CC(C)C[Si](CC(C)C)(OC)OC | |
| 204.385 | |
| 204.39 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2052
TSCA : Yes