accessibility menu, dialog, popup

UserName

H3COSiCH3H3CH3CCH3OCH3

CAS RN 17980-32-4

IUPAC Name: dimethoxybis(2-methylpropyl)silane
Synonyms: dimethoxybis(2-methylpropyl)silane
Molecular Weight (g/mol): 204.39
Molecular Formula: C10H24O2Si
InChi Key: NHYFIJRXGOQNFS-UHFFFAOYSA-N
SMILES: CO[Si](CC(C)C)(CC(C)C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results