missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyldimethoxysilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 72°C |
| Quantity | 5 mL |
| Formula Weight | 204.39 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diisobutyldimethoxysilane |
Safety and Handling
| EINECSNumber | (2)-2052 |
| TSCA | Yes |