missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate, SPEX CertiPrep™

Catalog No. S4150
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S4150 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S4150 Supplier SPEX CertiPrep Group LLC Supplier No. S4150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diisobutyl phthalate84-69-51000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 84-69-5
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00026480
InChI Key MGWAVDBGNNKXQV-UHFFFAOYSA-N
PubChem CID 6782
ChEBI CHEBI:79053
IUPAC Name 1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
SMILES CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Diisobutyl phthalate
Hazard Category Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only