missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyl phthalate, 99%, Thermo Scientific™
Specifications
Specifications
| Melting Point | -64°C |
| Density | 1.039 |
| Boiling Point | 327°C |
| Flash Point | 169°C (336°F) |
| Refractive Index | 1.49 |
| Quantity | 100 g |
| Beilstein | 2054802 |
| Formula Weight | 278.35 |
| Percent Purity | 99% |
| Chemical Name or Material | Diisobutyl phthalate |
Safety and Handling
| Hazard Category | H360Df |
| Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 201-553-2 |
| RTECSNumber | TI1225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only