missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 358730-88-8 |
| Flash Point | 109°C - Closed Cup |
| Physical Form | Neat |
| Linear Formula | C16D4H18O4 |
| MDL Number | MFCD01861606 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 282.37 |
| Grade | Analytical Standard |