missing translation for 'onlineSavingsMsg'
Learn More

Diisoamyl Disulfide 98.0+%, TCI America™

Catalog No. I008525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I008525ML Supplier TCI America Supplier No. I008525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2051-04-9
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD00026520
InChI Key MPYGLNNTOXLWOB-UHFFFAOYSA-N
Synonym Isoamyl Disulfide
PubChem CID 74915
IUPAC Name 3-methyl-1-(3-methylbutyldisulfanyl)butane
SMILES CC(C)CCSSCCC(C)C

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 mL
Formula Weight 206.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisoamyl Disulfide