missing translation for 'onlineSavingsMsg'
Learn More

Dihydrostreptomycin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014580
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014580 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014580 Supplier Merck Emd Millipore Supplier No. PHR1517500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dihydrostreptomycin sulfate is a semi-synthetic broad-spectrum aminoglycoside antibiotic produced commonly by the reduction of streptomycin. It is used primarily in the treatment of tuberculosis.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5490-27-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H41N7O12 · 3/2H2 SO4
MDL Number MFCD00070252
Synonym Dihydrostreptomycin sesquisulfate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 730.71
Grade Certified Reference Material