missing translation for 'onlineSavingsMsg'
Learn More

Dihydrolipoic Acid 97.0+%, TCI America™

Supplier:  TCI America D45861G

Quantity 1 g

 View more versions of this product

Catalog No. D45861G


Only null left

Description

Description

Specifications

Chemical Identifiers

462-20-4
208.334
IZFHEQBZOYJLPK-UHFFFAOYSA-N
421
6,8-bis(sulfanyl)octanoic acid
C8H16O2S2
MFCD00166974
6,8-Dimercapto-n-octanoic Acid
CHEBI:18047
C(CCC(=O)O)CC(CCS)S

Specifications

Dihydrolipoic Acid
Yellow
C8H16O2S2
MFCD00166974
IZFHEQBZOYJLPK-UHFFFAOYSA-N
6,8-bis(sulfanyl)octanoic acid
421
208.33
Liquid
462-20-4
175°C
1 g
6,8-Dimercapto-n-octanoic Acid
C(CCC(=O)O)CC(CCS)S
208.334
CHEBI:18047
≥97.0% (GC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RH0525000

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions