missing translation for 'onlineSavingsMsg'
Learn More

Dihydrolipoic Acid 97.0+%, TCI America™

Catalog No. p-7128355
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4586200MG 200 mg
D45861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4586200MG Supplier TCI America Supplier No. D4586200MG

Chemical Identifiers

CAS 462-20-4
Molecular Formula C8H16O2S2
Molecular Weight (g/mol) 208.334
MDL Number MFCD00166974
InChI Key IZFHEQBZOYJLPK-UHFFFAOYSA-N
Synonym 6,8-Dimercapto-n-octanoic Acid
PubChem CID 421
ChEBI CHEBI:18047
IUPAC Name 6,8-bis(sulfanyl)octanoic acid
SMILES C(CCC(=O)O)CC(CCS)S

Specifications

Color Yellow
Boiling Point 175°C
Quantity 200 mg
Formula Weight 208.33
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dihydrolipoic Acid
RTECSNumber RH0525000
TSCA No
Recommended Storage Freezer