missing translation for 'onlineSavingsMsg'
Learn More

Dihydrojasmonic Acid 98.0+%, TCI America™

Catalog No. D32251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32251G 1 g
D32255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32251G Supplier TCI America Supplier No. D32251G
Only null left

Chemical Identifiers

CAS 3572-64-3
Molecular Formula C12H20O3
Molecular Weight (g/mol) 212.29
MDL Number MFCD18781916
InChI Key PQEYTAGBXNEUQL-UHFFFAOYNA-N
Synonym 2-Amyl-3-(carboxymethyl)cyclopentanone, 2-Amyl-3-oxocyclopentaneacetic Acid, 3-(Carboxymethyl)-2-pentylcyclopentanone, 3-Oxo-2-pentylcyclopentaneacetic Acid
PubChem CID 107126
IUPAC Name 2-(3-oxo-2-pentylcyclopentyl)acetic acid
SMILES CCCCCC1C(CC(O)=O)CCC1=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 212.29
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Dihydrojasmonic Acid
TSCA Yes
Recommended Storage Freezer