accessibility menu, dialog, popup

UserName

CAS RN 3572-64-3

H3COOHO

CAS RN 3572-64-3

IUPAC Name: 2-(3-oxo-2-pentylcyclopentyl)acetic acid
Synonyms: (3-oxo-2-pentylcyclopentyl)acetic acid
Molecular Weight (g/mol): 212.29
Molecular Formula: C12H20O3
InChi Key: PQEYTAGBXNEUQL-UHFFFAOYNA-N
SMILES: CCCCCC1C(CC(O)=O)CCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results