missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min

Catalog No. p-7048952
Change view
Click to view available options
Quantity:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1217802 0.5 g
AA1217803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1217802 Supplier Thermo Scientific Chemicals Supplier No. 01217802
Only null left

CAS: 27057-71-2 | H2Cl6Os | 404.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27057-71-2
Molecular Formula H2Cl6Os
Molecular Weight (g/mol) 404.93
MDL Number MFCD00011329
InChI Key GCRLVKBHFZOVLQ-UHFFFAOYSA-H
Synonym Hydrogen hexachloroosmate(IV); Chloroosmic acid
IUPAC Name hexachloroosmium
SMILES Cl[Os](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Color Black
Linear Formula H2OsCl6·xH2O
Quantity 0.5 g
UN Number UN3260
Sensitivity Hygroscopic
Formula Weight 404.93 (Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 37% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroosmate(IV) hydrate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 248-192-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only