accessibility menu, dialog, popup

UserName

OsClClClClClClHH

CAS RN 27057-71-2

IUPAC Name: hexachloroosmium
Synonyms: hexachloroosmium
Molecular Weight (g/mol): 404.93
Molecular Formula: H2Cl6Os
InChi Key: GCRLVKBHFZOVLQ-UHFFFAOYSA-H
SMILES: Cl[Os](Cl)(Cl)(Cl)(Cl)Cl
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)