missing translation for 'onlineSavingsMsg'
Learn More

Dihydrofluorescein diacetate, 97%

Fluorescent dye for detecting intracellular oxidants | CAS: 35340-49-9 | C24H18O7 | 418.401 g/mol

$151.10 - $418.38

Chemical Identifiers

CAS 35340-49-9
Molecular Formula C24H18O7
Molecular Weight (g/mol) 418.401
MDL Number MFCD00005056
InChI Key YKSJJXGQHSESKB-UHFFFAOYSA-N
Synonym dihydrofluorescein diacetate, 2-3,6-diacetoxy-9h-xanthen-9-yl benzoic acid, diacetyldihydrofluorescein, 2-3,6-diacetoxy-9h-xanth-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid, dihydrofluorescein diacetate h2fda, 2-3,6-diacetyloxy-9h-xanthen-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid #
PubChem CID 629056
IUPAC Name 2-(3,6-diacetyloxy-9H-xanthen-9-yl)benzoic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C(C3=C(O2)C=C(C=C3)OC(=O)C)C4=CC=CC=C4C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65759MD
View Documents
Thermo Scientific Chemicals
J65759MD
250 mg
Each for $151.10
Only null left
 
AAJ6575903
View Documents
Thermo Scientific Chemicals
J6575903
1 g
Each for $418.38
Only null left
 
Description

Description

Fluorescent dye for detecting intracellular oxidants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35340-49-9
418.401
YKSJJXGQHSESKB-UHFFFAOYSA-N
629056
CC(=O)OC1=CC2=C(C=C1)C(C3=C(O2)C=C(C=C3)OC(=O)C)C4=CC=CC=C4C(=O)O
C24H18O7
MFCD00005056
dihydrofluorescein diacetate, 2-3,6-diacetoxy-9h-xanthen-9-yl benzoic acid, diacetyldihydrofluorescein, 2-3,6-diacetoxy-9h-xanth-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid, dihydrofluorescein diacetate h2fda, 2-3,6-diacetyloxy-9h-xanthen-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid #
2-(3,6-diacetyloxy-9H-xanthen-9-yl)benzoic acid

Specifications

35340-49-9
White to Yellow
MFCD00005056
364167
YKSJJXGQHSESKB-UHFFFAOYSA-N
2-(3,6-diacetyloxy-9H-xanthen-9-yl)benzoic acid
629056
97%
Dihydrofluorescein diacetate
213°C to 215°C
C24H18O7
250 mg
dihydrofluorescein diacetate, 2-3,6-diacetoxy-9h-xanthen-9-yl benzoic acid, diacetyldihydrofluorescein, 2-3,6-diacetoxy-9h-xanth-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid, dihydrofluorescein diacetate h2fda, 2-3,6-diacetyloxy-9h-xanthen-9-yl benzoic acid, 2-3,6-bis acetyloxy-9h-xanthen-9-yl benzoic acid #
CC(=O)OC1=CC2=C(C=C1)C(C3=C(O2)C=C(C=C3)OC(=O)C)C4=CC=CC=C4C(=O)O
418.401
418.39
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only