missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Dihydroethidium, Calbiochem™,

Catalog No. 30980025mg
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30980025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30980025MG Supplier MilliporeSigma™ Supplier No. 30980025MG
Only null left

A cell-permeable, chemically-reduced ethidium derivative

Chemical Identifiers

CAS 38483-26-0
Molecular Formula C21H21N3
Molecular Weight (g/mol) 315.42
InChI Key XYJODUBPWNZLML-UHFFFAOYSA-N
Synonym dihydroethidium, hydroethidine, 5-ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, 3,8-phenanthridinediamine, 5-ethyl-5,6-dihydro-6-phenyl, 3,8-phenanthridinediamine, 5-ethyl-5,6-dihydro-6-phenyl-, +-, dihydroethidium, bioreagent, suitable for fluorescence hpce, 2,7-diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, 3,8-diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine
PubChem CID 128682
IUPAC Name 5-ethyl-6-phenyl-6H-phenanthridine-3,8-diamine
SMILES CCN1C(C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N)C4=CC=CC=C4

Specifications

Color Tan
Quantity 25 mg
Solubility Information DMF or DMSO
Formula Weight 315.4
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material Dihydroethidium