missing translation for 'onlineSavingsMsg'
Learn More

Dihydroartemisinin 98.0+%, TCI America™

Catalog No. D37931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37931G 1 g
D37935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37931G Supplier TCI America Supplier No. D37931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71939-50-9
Molecular Formula C15H24O5
Molecular Weight (g/mol) 284.35
MDL Number MFCD00236467,MFCD00274495,MFCD00274495,MFCD28384156
InChI Key BJDCWCLMFKKGEE-KXTPALSWSA-N
PubChem CID 44264589
IUPAC Name (4S,5R,8S,9R,10S,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-ol
SMILES C[C@@H]1CC[C@H]2[C@@H](C)[C@@H](O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 284.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dihydroartemisinin
RTECSNumber KD4165550
TSCA No
Recommended Storage Refrigerator