accessibility menu, dialog, popup

UserName

CAS RN 71939-50-9

IUPAC Name: (4S,5R,8S,9R,10S,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-ol
Synonyms: artenimol
Molecular Weight (g/mol): 284.35
Molecular Formula: C15H24O5
InChi Key: BJDCWCLMFKKGEE-KXTPALSWSA-N
SMILES: C[C@@H]1CC[C@H]2[C@@H](C)[C@@H](O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results