missing translation for 'onlineSavingsMsg'
Learn More

Dihexylamine 98.0+%, TCI America™

Supplier:  TCI America D054725ML

 View more versions of this product

Catalog No. D054725ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dihexylamine
Yellow
194°C
MFCD00009521
2922
PXSXRABJBXYMFT-UHFFFAOYSA-N
N-hexylhexan-1-amine
8920
≥98.0% (GC)
143-16-8
9.9 (0.3g/L H2O soln. 25°C)
C12H27N
25 mL
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
CCCCCCNCCCCCC
185.355
185.36
Liquid

Chemical Identifiers

143-16-8
185.355
PXSXRABJBXYMFT-UHFFFAOYSA-N
8920
CCCCCCNCCCCCC
C12H27N
MFCD00009521
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
N-hexylhexan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3967

RTECSNumber : IH6600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions