missing translation for 'onlineSavingsMsg'
Learn More

Dihexylamine 98.0+%, TCI America™

Catalog No. p-7126278
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D054725ML 25 mL
D0547100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D054725ML Supplier TCI America Supplier No. D054725ML
Only null left

Chemical Identifiers

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC

Specifications

Color Yellow
pH 9.9 (0.3g/L H2O soln. 25°C)
Boiling Point 194°C
Quantity 25 mL
UN Number 2922
Formula Weight 185.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dihexylamine
EINECSNumber (2)-3967
RTECSNumber IH6600000
TSCA Yes