missing translation for 'onlineSavingsMsg'
Learn More

Dihexyl N,N-Diethylcarbamylmethylenephosphonate 90.0+%, TCI America™

Catalog No. D16635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16635G Supplier TCI America Supplier No. D16635G
Only null left

Chemical Identifiers

CAS 7369-66-6
Molecular Formula C18H38NO4P
Molecular Weight (g/mol) 363.48
MDL Number MFCD00015502
InChI Key MSULAAVSYZOTHM-UHFFFAOYSA-N
Synonym N,N-Diethylcarbamoylmethylphosphonic Acid Dihexyl Ester, Dihexyl N,N-Diethylcarbamoylmethylphosphonate, DHDECMP
PubChem CID 12865231
IUPAC Name dihexyl [(diethylcarbamoyl)methyl]phosphonate
SMILES CCCCCCOP(=O)(CC(=O)N(CC)CC)OCCCCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 363.48
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dihexyl N,N-Diethylcarbamylmethylenephosphonate [for Extraction of Lanthanides and Actinides]