accessibility menu, dialog, popup

UserName

H3COPOONH3CCH3OCH3

CAS RN 7369-66-6

IUPAC Name: dihexyl [(diethylcarbamoyl)methyl]phosphonate
Synonyms: dihexyl (diethylcarbamoyl)methylphosphonate
Molecular Weight (g/mol): 363.48
Molecular Formula: C18H38NO4P
InChi Key: MSULAAVSYZOTHM-UHFFFAOYSA-N
SMILES: CCCCCCOP(=O)(CC(=O)N(CC)CC)OCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results