missing translation for 'onlineSavingsMsg'
Learn More

Diheptyl phthalate, Analytical Standard, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014036
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014036 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014036 Supplier Merck Emd Millipore Supplier No. 002821ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 3648-21-3
Density 0.988 g/mL (at 25°C)
Linear Formula C6H4-1,2-[CO2(CH2)6 CH3]2
MDL Number MFCD00043680
Boiling Point 195°C (0.04 mmHg)
Synonym Phthalic acid diheptyl ester
Quantity 1 mL
Formula Weight 362.50
Percent Purity ≥97.0% (GC)
Grade Analytical Standard